Search
View additional product information for GeneChip™ Fluidics Station 450Dx v.2 - FAQs (00-0335)
3 product FAQs found
The following steps outline how to determine which step in the protocol you were on:
- Open Individual Station ID
- Under the steps, click the down arrow, and the step you were on should be highlighted
- After troubleshooting the error, resume at the step that was previously highlighted
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Please refer to pages 79-80 in the Affymetrix GeneChip Fluidics Station 450 User's Guide for AGCC (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/fs450_agcc_user_guide.pdf) for instructions on how to set up the dip switch.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
For long-term storage, we recommend running the shutdown process with water and running the shutdown process again with air. Remove the tubes from the wash buffer bottles and remove the peristaltic tubing. The fluidics station should now be ready for long-term storage.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.