GeneTitan™ MC Fast Scan Instrument, international (220V)
Applied Biosystems™

GeneTitan™ MC Fast Scan Instrument, international (220V)

The Applied Biosystems GeneTitan MC Fast Scan Instrument and proven GeneChip array using Axiom 2.0 assay are the first toRead more
Have Questions?
Catalog NumberQuantity
00-03731 Ea.
Catalog number 00-0373
Price (INR)
-
Quantity:
1 Ea.
The Applied Biosystems GeneTitan MC Fast Scan Instrument and proven GeneChip array using Axiom 2.0 assay are the first to provide a hands-free, automated solution for genome-wide SNP genotyping and copy number variation analysis in human and agrigenomics.

Features of the GeneTitan MC Fast Scan Instrument include:
• Fast—condenses hands-on processing time to as little as 30 minutes, images an array in less than five minutes, and operates unattended overnight
• Scalable—meets both medium- and high-throughput needs, enables fast time to data, and requires minimal manual intervention
• Flexible—supports genotyping studies on multiple array plate formats
• Accurate—enables high-quality, consistent data by processing multiple samples under identical conditions
• Adaptable—creates flexible workflows and sample registration via GeneChip Command Console Software

The GeneTitan MC Fast Scan Instrument automates array processing from target hybridization to data generation by combining a hybridization oven, fluidics processing, and state-of-the art imaging device into a single bench-top instrument. The imaging device in the instrument uses an external, high-intensity xenon lamp and dual excitation and emission filters to capture images from array plates.

The GeneTitan MC Fast Scan Instrument provides the highest throughput and laboratory productivity per technician by minimizing user intervention and allowing unattended, overnight processing of large numbers of samples in parallel. This unique degree of hands-free automation means your data will be consistently reproducible, your lab will be more productive, and you can spend less time acquiring and managing data and have more time for science.

To learn more about how the GeneTitan MC Fast Scan Instrument can maximize productivity and data reproducibility in your lab, please contact us today.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Dimensions55 x 33 x 26 in. (139.7 x 83.82 x 66 cm)
Quantity1 Ea.
For Use With (Application)Microarray Processing
Weight147 kg
Unit SizeEach

Frequently asked questions (FAQs)

What are the power specifications for the GeneTitan MC Instrument?

Specifications for the GeneTitan MC instrument can be found in the "Automated array processing with the GeneTitan family of instruments" data sheet (http://media.affymetrix.com/support/technical/datasheets/genetitan_family_instrument_datasheet.pdf).

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

Are there any service providers for the Axiom Genome-Wide BOS 1 Array Plate?

A number of service providers have experience running the Axiom Assay and the genotyping call algorithm that is used with the Axiom Genome-Wide BOS 1 Array Plate.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

What size are the files associated with the Axiom Genome-Wide BOS 1 Array Plate?

The average sample file size is 28 MB.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

What quality control tools are available for the Axiom Genome-Wide BOS 1 Array?

Dish QC (DQC) is the recommended QC metric for Axiom Genome-Wide Arrays in Genotyping Console Software. The default threshold is greater than or equal to 0.82 for each sample. It operates by measuring signal at a collection of sites in the genome that are known not to vary from one individual to the next. Because the metric monitors non-polymorphic locations, at each position it is known which of the two channels in the assay should contain signal and which should be just background. DQC is a measure of the extent to which the distribution of signal values is separated from background values, with 0 indicating no separation and 1 indicating perfect separation.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

How do I analyze the data generated by the Axiom Genome-Wide BOS 1 Array Plate?

Axiom Analysis Suite is used to analyze data from the Axiom Genome-Wide BOS 1 Array Plate.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.