eBioscience™ 10X RBC Lysis Buffer (Multi-species)
Invitrogen™
eBioscience™ 10X RBC Lysis Buffer (Multi-species)
This 10X RBC Lysis Buffer (Multi-species) is specially formulated for optimal lysis of erythrocytes in single-cell suspensions of peripheral bloodRead more
Have Questions?
Catalog NumberQuantity
00-4300-5450 mL
Catalog number 00-4300-54
Price (USD)
71.65
Online exclusive
96.00
Save 24.35
Each
Quantity:
50 mL
Price (USD)
71.65
Online exclusive
96.00
Save 24.35
Each
This 10X RBC Lysis Buffer (Multi-species) is specially formulated for optimal lysis of erythrocytes in single-cell suspensions of peripheral blood and hematopoietic tissues such as spleen. This buffer can be used for lysis of human, mouse, rat, canine, and non-human primate samples. 10X RBC Lysis Buffer (Multi-species) contains ammonium chloride, which lyses red blood cells with a minimal effect on lymphocytes when used as instructed.
Reactivity/Species
Canine, Human, Mouse, Non-human primate, Rat, Rhesus
Reported Application
Flow Cytometric Analysis, Functional Assays, Cell Culture
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatLiquid
Quantity50 mL
Volume (Metric)50 mL
Product LineeBioscience
Product TypeLysis Buffer
Unit SizeEach
Frequently asked questions (FAQs)
We've purchased the eBioscience 10X RBC Lysis Buffer (Multi-species) (Cat. No. 00-4300-54). Can we store the reconstituted solution at room temp or is it necessary to prepare the solution fresh before use?
The diluted 10X RBC Lysis Buffer should be stable for about 1 month when stored at room temperature. The product will be more stable as a 10X concentrate. We recommend making enough of the 1X solution to last 3-4 weeks.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.