Product Image
Invitrogen™
eBioscience™ 20X TBS Wash Buffer for IHC/ICC
The 20X TBS Wash Buffer for IHC/ICC is for use in immunohistochemistry and immunocytochemistry protocols which require washing to removeRead more
Have Questions?
Catalog NumberQuantity
00-4954-56100 mL
Catalog number 00-4954-56
Price (CLP)
136.824
Each
Quantity:
100 mL
Price (CLP)
136.824
Each
The 20X TBS Wash Buffer for IHC/ICC is for use in immunohistochemistry and immunocytochemistry protocols which require washing to remove non-specific binding. This wash buffer is recommended for eFluor™ Nanocrystal conjugated-antibodies following antibody incubation. The TBS Wash Buffer is also compatible with organic dye-conjugated antibodies as well as unconjugated antibodies that may be used in two and three step staining protocols. The buffer is supplied as a 20X stock solution that should be diluted to 1X with distilled water.
Reported Application
Immunohistochemical Staining, Immunocytochemistry
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormLiquid
Product TypeWash Buffer
For Use With (Application)Immunocytochemistry (ICC), Immunohistochemistry (IHC)
Product LineeBioscience
Quantity100 mL
Unit SizeEach