Search
View additional product information for Fluoromount-G™ Mounting Medium, with DAPI - FAQs (00-4959-52)
1 product FAQs found
Fluoromount-G Mounting Medium, with DAPI (Cat. No. 00-4959-52) is best used when the thickness of the specimen is less than 10 µm.