Product Image
Invitrogen™
eBioscience™ Cell Stimulation Cocktail (plus protein transport inhibitors) (500X)
The Cell Stimulation Cocktail (plus protein transport inhibitors) (500X) is a cocktail of phorbol 12-myristate 13-acetate (PMA), ionomycin, brefeldin ALeia mais
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoQuantity
00-4975-93100 μL
00-4975-034 x 100 μL
Número do catálogo 00-4975-93
Preço (BRL)
1.171,91
Each
Quantity:
100 μL
Preço (BRL)
1.171,91
Each
The Cell Stimulation Cocktail (plus protein transport inhibitors) (500X) is a cocktail of phorbol 12-myristate 13-acetate (PMA), ionomycin, brefeldin A and monensin. Treatment with PMA and ionomycin is sufficient to induce activation of many cell types to produce cytokines. Brefeldin A and Monensin result in the accumulation of secreted proteins in the endoplasmic reticulum and Golgi apparatus. Thus, the Cell Stimulation Cocktail (plus protein transport inhibitors) (500X) can be used for induction and subsequent intracellular detection of cytokines and other secreted proteins in both in vitro-cultured and ex vivo cells.
For an unstimulated and vehicle control, see the Protein Transport Inhibitor Cocktail (500X) (cat. 00-4980).
Reported Application
Flow Cytometric Analysis, Functional Assays, Cell Culture
For Research Use Only. Not for use in diagnostic procedures.
Especificações
Quantity100 μL
For Use With (Application)Flow Cytometry, Functional Assays, Cell Culture
Product LineeBioscience
Product TypeCell Stimulation Cocktail
Unit SizeEach