eBioscience™ Lipopolysaccharide (LPS) Solution (500X), 100 μL - FAQs
eBioscience™ Lipopolysaccharide (LPS) Solution (500X), 100 μL - FAQs
View additional product information for eBioscience™ Lipopolysaccharide (LPS) Solution (500X) - FAQs (00-4976-93, 00-4976-03)
1 product FAQs found
Can I freeze thaw eBioscience Lipopolysaccharide (LPS) Solution (500X)?
eBioscience Lipopolysaccharide (LPS) Solution (500X) has been tested under conditions where no change in performance was observed after 20 freeze-thaw cycles. For optimal performance, we recommend preparing smaller aliquots to minimize the number of freeze-thaw cycles.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.