Product Image
Invitrogen™
eBioscience™ Concanavalin A (Con A) Solution (500X)
The Concanavalin A (Con A) Solution (500X) is a ready-to-use solution of Con A in aqueous buffer. Con A isRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
00-4978-93100 μL
00-4978-034 x 100 μL
Catalog number 00-4978-93
Price (KRW)
74,000
Online offer
Ends: 30-Sep-2026
87,000
Save 13,000 (15%)
Each
Quantity:
100 μL
Price (KRW)
74,000
Online offer
Ends: 30-Sep-2026
87,000
Save 13,000 (15%)
Each
The Concanavalin A (Con A) Solution (500X) is a ready-to-use solution of Con A in aqueous buffer. Con A is isolated from the Jack bean, Canavlia ensiformis. It is a lectin that binds alpha-D-glucose and alpha-D-mannose moieties found in various glycoproteins, glycolipids, and sugars and is a potent leukocyte mitogen. This reagent is intended for use in in vitro activation of human and mouse leukocytes.
Reported Application
Functional Assays
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatVial
Product LineeBioscience
Protein SubtypeLectins
Quantity100 μL
SourceCanavlia Ensiformis
FormSolution
Unit SizeEach