Applied Biosystems™

Eureka™ Genotyping Services, Catalog Panels

Catalog number: 000.900
Applied Biosystems™

Eureka™ Genotyping Services, Catalog Panels

Catalog number: 000.900
Catalog Number
000.900
also known as 000900
Unit Size
1 each
Price (USD)
Catalog NumberUnit SizePrice (USD)
000.900
also known as 000900
1 eachContact Us ›
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Eureka™ Genotyping Services™ Catalog Panels
Our service lab has genotyped more than one million samples and called about 650 billion genotypes, enabling our customers to complete their genotyping studies quickly and affordably. Our staff uses expert knowledge of best practices to process and analyze your samples, manage your Eureka Genotyping Services project from start to finish, and meet your crucial deadlines. Descriptions of the variety of Eureka Genotyping Services offered are listed below.

Eureka Catalog Panel Genotyping Services for Agrigenomics
Our Eureka Catalog Panel Genotyping Services work collaboratively with customers, providing a personal touch to achieve your research goals. Using state-of-the-art technology, you can fast-track your genomic selection program.

Eureka™ myDesign™ Panel Genotyping Services for Agrigenomics
Our Eureka myDesign Panel Genotyping Services enable you to customize your genotyping panel containing 100 to 3,000 markers. They will be processed by our Research Services Laboratory with the same high-quality and fast turnaround time we provide for our Eureka catalog panels.

For any of the above services, Please inquire for more information.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Array
Genotyping
Assay Service Type
Genotyping Services
Species
Human

Figures

Frequently asked questions (FAQs)

Citations & References