Thermo Scientific™

Dysprosium pieces, 99.9% (REO), Thermo Scientific™

50 g, Each
Thermo Scientific™

Dysprosium pieces, 99.9% (REO), Thermo Scientific™

50 g, Each
Quantity
Catalog number: 000106.18
also known as 000106-18
Price (USD)
587.00 / Each
Your Price:
Quantity
-

Chemical Identifiers

CAS
7429-91-6
IUPAC Name
dysprosium
Molecular Formula
Dy
InChI Key
KBQHZAAAGSGFKK-UHFFFAOYSA-N
SMILES
[Dy]
Total Rare Earth Oxide Impurities
0.1% max.

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
  • Packaged under argon
RUO – Research Use Only

Figures