Indium pieces, 99.99% (metals basis)
Indium pieces, 99.99% (metals basis)
Indium pieces, 99.99% (metals basis)
Thermo Scientific Chemicals

Indium pieces, 99.99% (metals basis)

CAS: 7440-74-6 | In | 114.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 000149.09
also known as 000149-09
Price (USD)
80.65
Online Exclusive
89.80
Save 9.15 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
80.65
Online Exclusive
89.80
Save 9.15 (10%)
Each
Chemical Identifiers
CAS7440-74-6
IUPAC Nameindium
Molecular FormulaIn
InChI KeyAPFVFJFRJDLVQX-UHFFFAOYSA-N
SMILES[In]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only