Phosphorus lump, red, Puratronic™, 99.999+% (metals basis), Thermo Scientific Chemicals
Phosphorus lump, red, Puratronic™, 99.999+% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phosphorus lump, red, Puratronic™, 99.999+% (metals basis), Thermo Scientific Chemicals

 
Have Questions?
Quantity:
50 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 000248.18
also known as 000248-18
Price (USD)/ Each
604.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
711.00 
Save 106.35 (15%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
604.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
711.00 
Save 106.35 (15%)
Add to cart
Phosphorus lump, red, Puratronic™, 99.999+% (metals basis), Thermo Scientific Chemicals
Catalog number000248.18
Price (USD)/ Each
604.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
711.00 
Save 106.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namephosphorus
Molecular FormulaP
InChI KeyOAICVXFJPJFONN-UHFFFAOYSA-N
SMILES[P]
Molecular Weight (g/mol)30.97
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only