Titanium wire, 0.5mm (0.02in) dia, 99.98% (metals basis)
Titanium wire, 0.5mm (0.02in) dia, 99.98% (metals basis)
Titanium wire, 0.5mm (0.02in) dia, 99.98% (metals basis)
Thermo Scientific Chemicals

Titanium wire, 0.5mm (0.02in) dia, 99.98% (metals basis)

Form: approx. 0.88g/m | CAS: 7440-32-6 | Ti | 47.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 m
5 m
Catalog number 000362.G1
also known as 000362-G1
Price (EUR)
184,00
Each
Quantity:
1 m
Request bulk or custom format
Price (EUR)
184,00
Each
Chemical Identifiers
CAS7440-32-6
IUPAC Nametitanium
Molecular FormulaTi
InChI KeyRTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES[Ti]
View more
SpecificationsSpecification SheetSpecification Sheet
FormWire, 0.5mm (0.02in) diameter.
Assay from Supplier's CofA≥99.98% (metals basis)
Appearance (Color)Silvery
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only