Chromium pieces, 3-12mm (0.12-0.47in), 99.2% (metals basis)
Chromium pieces, 3-12mm (0.12-0.47in), 99.2% (metals basis)
Chromium pieces, 3-12mm (0.12-0.47in), 99.2% (metals basis)
Thermo Scientific Chemicals

Chromium pieces, 3-12mm (0.12-0.47in), 99.2% (metals basis)

CAS: 7440-47-3 | Cr | 52.00 g/mol
Quantity:
100 g
500 g
Catalog number 000472.22
also known as 000472-22
Price (JPY)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS7440-47-3
IUPAC Namechromium
Molecular FormulaCr
InChI KeyVYZAMTAEIAYCRO-UHFFFAOYSA-N
SMILES[Cr]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPieces, 3-12mm
Total Metal Impurities≤0.8%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only