Tungsten slug, 17.5mm (0.689in) dia x 6.4mm (0.25in) length, 99.9% (metals basis)
Tungsten slug, 17.5mm (0.689in) dia x 6.4mm (0.25in) length, 99.9% (metals basis)
Tungsten slug, 17.5mm (0.689in) dia x 6.4mm (0.25in) length, 99.9% (metals basis)
Tungsten slug, 17.5mm (0.689in) dia x 6.4mm (0.25in) length, 99.9% (metals basis)
Thermo Scientific Chemicals

Tungsten slug, 17.5mm (0.689in) dia x 6.4mm (0.25in) length, 99.9% (metals basis)

CAS: 7440-33-7 | W | 183.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 Pc.
1 Pc.
Catalog number 000525.KI
also known as 000525-KI
Price (USD)/ Each
368.65
Special offer
Online exclusive
433.00
Save 64.35 (15%)
Add to cart
Quantity:
4 Pc.
Request bulk or custom format
Price (USD)/ Each
368.65
Special offer
Online exclusive
433.00
Save 64.35 (15%)
Add to cart
Chemical Identifiers
IUPAC Nametungsten
Molecular FormulaW
InChI KeyWFKWXMTUELFFGS-UHFFFAOYSA-N
SMILES[W]
Molecular Weight (g/mol)183.84
View more
SpecificationsSpecification SheetSpecification Sheet
FormSlug, 17.5mm (0.689in) diameter.
Assay from Supplier's CofA≥99.9% (metals basis)
Appearance (Color)Gray
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only