Copper powder, -100 mesh, 99%
Copper powder, -100 mesh, 99%
Copper powder, -100 mesh, 99%
Thermo Scientific Chemicals

Copper powder, -100 mesh, 99%

CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Catalog number 000584.A3
also known as 000584-A3
Price (EUR)
227,65
Online exclusive
253,00
Save 25,35 (10%)
Each
Quantity:
2 kg
Request bulk or custom format
Price (EUR)
227,65
Online exclusive
253,00
Save 25,35 (10%)
Each
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Copper
FormPowder, -100 mesh
Assay from Suppliers CofACopper: ≥99.0%
Particle Size-100 mesh
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only