Zinc wire, 0.25mm (0.01in) dia, 99.99+% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Zinc wire, 0.25mm (0.01in) dia, 99.99+% (metals basis), Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 m
1 m
Catalog number 000648.G5
also known as 000648-G5
Price (USD)/ Each
139.00
-
Add to cart
Quantity:
5 m
Request bulk or custom format
Price (USD)/ Each
139.00
Add to cart
Zinc wire, 0.25mm (0.01in) dia, 99.99+% (metals basis), Thermo Scientific Chemicals
Catalog number000648.G5
Price (USD)/ Each
139.00
-
Add to cart
Chemical Identifiers
CAS7440-66-6
IUPAC Namezinc
Molecular FormulaZn
InChI KeyHCHKCACWOHOZIP-UHFFFAOYSA-N
SMILES[Zn]
View more
SpecificationsSpecification SheetSpecification Sheet
FormWire, 0.25 mm diameter
Total Metal Impurities<0.01%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only