Lead foil, 1.6mm (0.063in) thick, 30x30cm, 99+% (metals basis)
Lead foil, 1.6mm (0.063in) thick, 30x30cm, 99+% (metals basis)
Lead foil, 1.6mm (0.063in) thick, 30x30cm, 99+% (metals basis)
Thermo Scientific Chemicals

Lead foil, 1.6mm (0.063in) thick, 30x30cm, 99+% (metals basis)

Form: Approx.1630g/30 x 30cm | CAS: 7439-92-1 | Pb | 207.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 Pc.
5 Pc.
Catalog number 000708.KF
also known as 000708-KF
Price (USD)
250.65
Online Exclusive
278.00
Save 27.35 (10%)
Each
Quantity:
1 Pc.
Request bulk or custom format
Price (USD)
250.65
Online Exclusive
278.00
Save 27.35 (10%)
Each
Chemical Identifiers
CAS7439-92-1
IUPAC Namelead
Molecular FormulaPb
InChI KeyWABPQHHGFIMREM-UHFFFAOYSA-N
SMILES[Pb]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil, 1.6mm (0.063in) thick.
Assay from Supplier's CofA>99% (metals basis)
Appearance (Color)Gray
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only