Gold wire, 0.25mm (0.01in) dia, 99.9% (metals basis)
Gold wire, 0.25mm (0.01in) dia, 99.9% (metals basis)
Gold wire, 0.25mm (0.01in) dia, 99.9% (metals basis)
Thermo Scientific Chemicals

Gold wire, 0.25mm (0.01in) dia, 99.9% (metals basis)

CAS: 7440-57-5 | Au | 196.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 m
5 m
Catalog number 000725.G1
also known as 000725-G1
Price (EUR)
-
Quantity:
1 m
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Specifications
Assay Percent Notes(metals basis)
Assay Percent Range99.9% (metals basis)
CAS7440-57-5
ChEBICHEBI:30050
Chemical Name or MaterialGold wire
EINECS Number231-165-9
IUPAC Namegold
InChI KeyPCHJSUWPFVWCPO-UHFFFAOYSA-N
MDL NumberMFCD00003436
Molecular FormulaAu
Name Note0.25mm (0.01 in.) dia.
PubChem CID23985
Recommended StorageAmbient temperatures
SMILES[Au]
Synonymcolloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
TSCAYes
Molecular Weight (g/mol)196.97
Percent Purity99.9%
Quantity1 m
Unit SizeEach