Bromine liquid, 99.8%, Thermo Scientific Chemicals
Bromine liquid, 99.8%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bromine liquid, 99.8%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 x 1 kg
25 g
250 g
1 kg
Catalog number 000905.D9
also known as 000905-D9
Price (USD)/ Each
594.00
-
Add to cart
Quantity:
5 x 1 kg
Request bulk or custom format
Price (USD)/ Each
594.00
Add to cart
Bromine liquid, 99.8%, Thermo Scientific Chemicals
Catalog number000905.D9
Price (USD)/ Each
594.00
-
Add to cart
Chemical Identifiers
IUPAC Namebromide
Molecular FormulaBr
InChI KeyCPELXLSAUQHCOX-UHFFFAOYSA-M
SMILES[Br-]
Molecular Weight (g/mol)79.91
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Dark red to very dark red or red-brown or dark red-browm
Assay (unspecified)≥99.80%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only