Product Image

AEC Solution (IHC)

This Single Solution utilizes aminoethyl carbazole (AEC), an alcohol soluble chromogen, and generates a reddish deposit at the reaction siteRead more
Have Questions?
Catalog NumberQuantity
0011221,000 slides
Catalog number 001122
Price (USD)
548.00
Each
Quantity:
1,000 slides
Price (USD)
548.00
Each
This Single Solution utilizes aminoethyl carbazole (AEC), an alcohol soluble chromogen, and generates a reddish deposit at the reaction site on tissue or cell preparations. Single Solution AEC is suitable for use with manual or automated staining systems, and is provided as a liquid in a single bottle that can be stored at -8°C.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialHRP (Horseradish Peroxidase) Substrate
Recommended StorageStore in refrigerator (2–8°C).

Shelf life: 18 months from date of manufacture
Quantity1,000 slides
Unit SizeEach