BCIP/NBT KIT

Catalog number: 002209

Novex™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The BCIP/NBT Kit may be used in alkaline phosphatase (AP)-based immunoblotting procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: Colorimetric
Product Size: 1,000 mL
Substrate: BCIP⁄NBT
Substrate Properties: Chemical Substrate
Substrate Type: AP (Alkaline Phosphatase) Substrate
Shipping Condition: Wet Ice

Contents & storage

Store at 2-8°C

Documents

Manuals & protocols

Recommended products