Digest-All Kit

Catalog number: 003006

Novex™  Related applications: Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Enzymatic digestion may be required to reveal antigenic sites in formalin-fixed, paraffinembedded tissue. However, optimal digestion conditions vary greatly depending on the fixation/antigen combination. The Digest-All has a flexible and standardized system for tissue digestion. The degree of digestion is based on a standardized 10 minute ncubation at 37°C. The trypsin solution is supplied with diluent so that different trypsin concentrations can be made to generate a range of digestion activity between that of Ficin and Pepsin. Digest-All™ 1, 2 and 3 are stable for 12 months. All components are in liquid form.
For In Vitro Diagnostic Use.

Specifications

Reagent Type: Antigen Retrieval Reagent
Product Line: Digest-All™
Product Size: 48 mL
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products