Ready-to-use diluent with BSA (for stabilizing the antibody) and preservative.
Have Questions?
Change view
Catalog Number
Quantity
003118
250 mL
003218
500 mL
2 Options
Catalog number 003118
Price (USD)
151.65
Online exclusive
167.00
Save 15.35
Each
Quantity:
250 mL
Price (USD)
151.65
Online exclusive
167.00
Save 15.35
Each
Ready-to-use diluent with BSA (for stabilizing the antibody) and preservative.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormLiquid
Product TypeAntibody Diluent
Shipping ConditionWet Ice
Quantity250 mL
Unit SizeEach
Contents & Storage
Store in refrigerator at 2°C to 8°C
Shelf life: 18 months from date of manufacture
Citations & References (1)
Citations & References
Abstract
Lysyl oxidase-related protein-1 promotes tumor fibrosis and tumor progression in vivo.
Authors:Akiri G, Sabo E, Dafni H, Vadasz Z, Kartvelishvily Y, Gan N, Kessler O, Cohen T, Resnick M, Neeman M, Neufeld G,
Journal:Cancer Res
PubMed ID:12670920
The lysyl oxidase gene family members function as extracellular matrix modulating enzymes. We have found that another member of this family, lysyl oxidase related protein-1 (LOR-1), is highly expressed in metastatic breast cancer-derived cell lines but not in the nonmetastatic estrogen-dependent MCF-7 cells. Furthermore, LOR-1 expression in periductal tumor cells ... More