DRI™ Ethyl Alcohol Drugs of Abuse Assays
Actual product may vary
DRI™ Ethyl Alcohol Drugs of Abuse Assays
DRI™ Ethyl Alcohol Drugs of Abuse Assays
Thermo Scientific™
DRI™ Ethyl Alcohol Drugs of Abuse Assays
Achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
00371 x 100 mL
00381 x 500 mL
100163973 x 18 mL
Catalog number 0037
Price (USD)
-
Quantity:
1 x 100 mL
Achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation with Thermo Scientific™ DRI™ Ethyl Alcohol Drugs of Abuse Assays, which offer applications for an array of clinical chemistry analyzers. These liquid, ready-to-use assays have been field-tested on a wide range of analyzers. They are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Control SetsDRI Ethyl Alcohol Control Set
DescriptionDRI Ethyl Alcohol Drugs of Abuse Assays
Quantity1 x 100 mL
Sample TypePlasma, Serum, Urine
TypeReagent
Detectable AnalytesAlcohol
DoA CalibratorsEthyl Alcohol Calibrator
Unit SizeEach
Contents & Storage
2°C to 8°C