ZipChip™ Assay Kits
Actual product may vary
ZipChip™ Assay Kits
ZipChip™ Assay Kits
ZipChip™ Assay Kits
Thermo Scientific™
ZipChip™ Assay Kits
Reduce error and enjoy the convenience of prepackaged ZipChip™ Assay Kits, which are for use on the ZipChip Interface for analysis of biological samples by MS.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
00950-01-00500Metabolites Assay Kit
00950-01-00501Peptide Assay Kit
00950-01-00502Intact Protein Assay Kit
Catalog number 00950-01-00500
Price (USD)
-
Type:
Metabolites Assay Kit
Reduce error and enjoy the convenience of prepackaged ZipChip™ Assay Kits. These kits are for use with the ZipChip Interface, with or without the ZipChip Autosampler. Each ZipChip Assay Kit contains 5 bottles of background electrolyte and sample diluents, which is sufficient for 500 sample injections. A bottle can be directly loaded into ZipChip Autosampler for automated use and matching to a 96-well plate run. Different kits have been optimized for peptides, intact antibody, and metabolites analyses.
Three types of pre-packed ZipChip Assay Kits are customized for analyses of intact proteins, pepetides, and metabolites
  • Total of 500-sample capacity per kit
  • 5 individual bottles with 100 sample capacity each (designed to match to 96-well plate sampling) can be directly loaded into ZipChip autosampler
Specifications
For Use With (Equipment)ZipChip Interface for Mass Spectrometry
IncludesBackground electrolyte and sample diluents optimized for metabolite analysis
TypeMetabolites Assay Kit
Unit SizeEach