Anti-Clumping Agent - FAQs
View additional product information for Anti-Clumping Agent - FAQs (010057AE)
1 product FAQs found
How much Anti-clumping agent should I use?
Use anti-clumping agent at a dilution of 1:250 to 1:1000 in culture medium. Test for impact on the cell culture and reduce concentration as the cell clumping is reduced.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.