I am performing flow cytometry. My compensation matrix has values over 100. That's not possible is it?
It is possible. But if you have a lot of values over 100, you probably need to look at the voltages that you are using for your data collection.