Search
View additional product information for Sorvall™ High Performance Centrifuge Bottle - FAQs (010-1456, 010-1455, 010-1459, 010-1458)
1 product FAQs found
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.