Bis(benzonitrile)dichloroplatinum(II), Pt 40% min
Bis(benzonitrile)dichloroplatinum(II), Pt 40% min
Bis(benzonitrile)dichloroplatinum(II), Pt 40% min
Thermo Scientific Chemicals

Bis(benzonitrile)dichloroplatinum(II), Pt 40% min

CAS: 14873-63-3 | C14H10Cl2N2Pt | 472.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
1 g
5 g
Catalog number 010004.02
also known as 010004-02
Price (EUR)
117,00
Each
Quantity:
0.5 g
Request bulk or custom format
Price (EUR)
117,00
Each
Chemical Identifiers
CAS14873-63-3
IUPAC Namebis(benzonitrile); dichloroplatinumbis(ylium)
Molecular FormulaC14H10Cl2N2Pt
InChI KeyFMBJZWFXNZMJPC-UHFFFAOYSA-L
SMILESCl[Pt++]Cl.N#CC1=CC=CC=C1.N#CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Pt 40% min
Bis(benzonitrile)dichloroplatinum(II) is used as a catalyst for asymmetric hydroformylation reactions, allylation reactions, cyclopropanation reactions, hydrosilylation reactions and carbene insertion into O-H bonds of alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only