Thermo Scientific™

ProPac™ HIC-10 HPLC Columns

Catalog number: 01005-259270
Thermo Scientific™

ProPac™ HIC-10 HPLC Columns

Catalog number: 01005-259270

Achieve high-resolution separation of proteins and peptides under nondenaturing conditions with Thermo Scientific™ ProPac™ HIC-10 LC Columns. Developed with multi-functional ligand attachments, these silica-based columns provide exceptional hydrolytic stability under the highly aqueous conditions used in HIC.

 
Catalog Number
01005-259270
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
20 mm
Length (Metric)
250 mm
Price (USD)
Availability
***
Full specifications
Quantity
-
Column FormatPreparative Column, Standard
Diameter (Metric)20 mm
Length (Metric)250 mm
Max. Pressure3500 psi (240 bar)
Particle Size5 μm
pH2.5 to 7.5
Pore Size300 Å
Surface Area100 m²/g
Temperature40°C
Stationary PhaseAmide/Ethyl
Product LineProPac
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeHydrophobic Interaction
EndcappedNo
PhaseHydrophobic Interaction
MaterialStainless Steel
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
2.1 mm
Length (Metric)
100 mm
Price (USD)
789.00
Your Price:
Availability
***
Full specifications
Quantity
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Length (Metric)100 mm
Max. Pressure3500 psi (240 bar)
Particle Size5 μm
pH2.5 to 7.5
Pore Size300 Å
Surface Area100 m²/g
Temperature40°C
Stationary PhaseAmide/Ethyl
Product LineProPac
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeHydrophobic Interaction
PhaseHydrophobic Interaction
MaterialStainless Steel
TypeAnalytical Column
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
10 mm
Length (Metric)
100 mm
Price (USD)
Availability
***
Full specifications
Quantity
-
Column FormatPreparative Column, Standard
Diameter (Metric)10 mm
Length (Metric)100 mm
Max. Pressure3500 psi (240 bar)
Particle Size5 μm
pH2.5 to 7.5
Pore Size300 Å
Surface Area100 m²/g
Temperature40°C
Stationary PhaseAmide/Ethyl
Product LineAcclaim
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeHydrophobic Interaction
EndcappedNo
PhaseHydrophobic Interaction
MaterialStainless Steel
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
4.6 mm
Length (Metric)
100 mm
Price (USD)
789.00
Your Price:
Availability
***
Full specifications
Quantity
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Length (Metric)100 mm
Max. Pressure3500 psi (240 bar)
Particle Size5 μm
pH2.5 to 7.5
Pore Size300 Å
Surface Area100 m²/g
Temperature40°C
USP TypeL1
Stationary PhaseAmide/Ethyl
Product LineProPac
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeHydrophobic Interaction
PhaseHydrophobic Interaction
MaterialStainless Steel
TypeAnalytical Column
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
7.8 mm
Length (Metric)
75 mm
Price (USD)
1,407.00
Your Price:
Availability
***
Full specifications
Quantity
Column FormatAnalytical Column
Diameter (Metric)7.8 mm
Length (Metric)75 mm
Max. Pressure3500 psi (240 bar)
Particle Size5 μm
pH2.5 to 7.5
Pore Size300 Å
Surface Area100 m²/g
Temperature40°C
Stationary PhaseAmide/Ethyl
Product LineProPac
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeHydrophobic Interaction
PhaseHydrophobic Interaction
MaterialStainless Steel
TypeAnalytical Column
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
4.6 mm
Length (Metric)
250 mm
Price (USD)
1,265.00
Your Price:
Availability
***
Full specifications
Quantity
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Length (Metric)250 mm
Max. Pressure3500 psi (240 bar)
Particle Size5 μm
pH2.5 to 7.5
Pore Size300 Å
Surface Area100 m²/g
Temperature40°C
Stationary PhaseAmide/Ethyl
Product LineProPac
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeHydrophobic Interaction
PhaseHydrophobic Interaction
MaterialStainless Steel
TypeAnalytical Column
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
10 mm
Length (Metric)
150 mm
Price (USD)
Availability
***
Full specifications
Quantity
-
Column FormatPreparative Column, Standard
Diameter (Metric)10 mm
Length (Metric)150 mm
Max. Pressure3500 psi (240 bar)
Particle Size5 μm
pH2.5 to 7.5
Pore Size300 Å
Surface Area100 m²/g
Temperature40°C
Stationary PhaseAmide/Ethyl
Product LineProPac
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeHydrophobic Interaction
PhaseHydrophobic Interaction
MaterialStainless Steel
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
22 mm
Length (Metric)
250 mm
Price (USD)
Availability
***
Full specifications
Quantity
-
Column FormatPreparative Column, Standard
Diameter (Metric)22 mm
Length (Metric)250 mm
Max. Pressure3500 psi (240 bar)
Particle Size5 μm
pH2.5 to 7.5
Pore Size300 Å
Surface Area100 m²/g
Temperature40°C
Stationary PhaseAmide/Ethyl
Product LineProPac
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeHydrophobic Interaction
PhaseHydrophobic Interaction
MaterialStainless Steel
Unit SizeEach
Showing 8 of 8
Catalog NumberSpecificationsUnit SizeParticle SizeDiameter (Metric)Length (Metric)Price (USD)AvailabilityQuantity
01005-259270Full specifications
Each5 μm20 mm250 mmRequest A Quote***-
Column FormatPreparative Column, Standard
Diameter (Metric)20 mm
Length (Metric)250 mm
Max. Pressure3500 psi (240 bar)
Particle Size5 μm
pH2.5 to 7.5
Pore Size300 Å
Surface Area100 m²/g
Temperature40°C
Stationary PhaseAmide/Ethyl
Product LineProPac
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeHydrophobic Interaction
EndcappedNo
PhaseHydrophobic Interaction
MaterialStainless Steel
Unit SizeEach
063653Full specifications
Each5 μm2.1 mm100 mm
789.00
Your Price:
***
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Length (Metric)100 mm
Max. Pressure3500 psi (240 bar)
Particle Size5 μm
pH2.5 to 7.5
Pore Size300 Å
Surface Area100 m²/g
Temperature40°C
Stationary PhaseAmide/Ethyl
Product LineProPac
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeHydrophobic Interaction
PhaseHydrophobic Interaction
MaterialStainless Steel
TypeAnalytical Column
Unit SizeEach
01005-109070Full specifications
Each5 μm10 mm100 mmRequest A Quote***-
Column FormatPreparative Column, Standard
Diameter (Metric)10 mm
Length (Metric)100 mm
Max. Pressure3500 psi (240 bar)
Particle Size5 μm
pH2.5 to 7.5
Pore Size300 Å
Surface Area100 m²/g
Temperature40°C
Stationary PhaseAmide/Ethyl
Product LineAcclaim
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeHydrophobic Interaction
EndcappedNo
PhaseHydrophobic Interaction
MaterialStainless Steel
Unit SizeEach
063655Full specifications
Each5 μm4.6 mm100 mm
789.00
Your Price:
***
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Length (Metric)100 mm
Max. Pressure3500 psi (240 bar)
Particle Size5 μm
pH2.5 to 7.5
Pore Size300 Å
Surface Area100 m²/g
Temperature40°C
USP TypeL1
Stationary PhaseAmide/Ethyl
Product LineProPac
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeHydrophobic Interaction
PhaseHydrophobic Interaction
MaterialStainless Steel
TypeAnalytical Column
Unit SizeEach
063665Full specifications
Each5 μm7.8 mm75 mm
1,407.00
Your Price:
***
Column FormatAnalytical Column
Diameter (Metric)7.8 mm
Length (Metric)75 mm
Max. Pressure3500 psi (240 bar)
Particle Size5 μm
pH2.5 to 7.5
Pore Size300 Å
Surface Area100 m²/g
Temperature40°C
Stationary PhaseAmide/Ethyl
Product LineProPac
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeHydrophobic Interaction
PhaseHydrophobic Interaction
MaterialStainless Steel
TypeAnalytical Column
Unit SizeEach
074197Full specifications
Each5 μm4.6 mm250 mm
1,265.00
Your Price:
***
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Length (Metric)250 mm
Max. Pressure3500 psi (240 bar)
Particle Size5 μm
pH2.5 to 7.5
Pore Size300 Å
Surface Area100 m²/g
Temperature40°C
Stationary PhaseAmide/Ethyl
Product LineProPac
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeHydrophobic Interaction
PhaseHydrophobic Interaction
MaterialStainless Steel
TypeAnalytical Column
Unit SizeEach
SP6800Full specifications
Each5 μm10 mm150 mmRequest A Quote***-
Column FormatPreparative Column, Standard
Diameter (Metric)10 mm
Length (Metric)150 mm
Max. Pressure3500 psi (240 bar)
Particle Size5 μm
pH2.5 to 7.5
Pore Size300 Å
Surface Area100 m²/g
Temperature40°C
Stationary PhaseAmide/Ethyl
Product LineProPac
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeHydrophobic Interaction
PhaseHydrophobic Interaction
MaterialStainless Steel
Unit SizeEach
SP6869Full specifications
Each5 μm22 mm250 mmRequest A Quote***-
Column FormatPreparative Column, Standard
Diameter (Metric)22 mm
Length (Metric)250 mm
Max. Pressure3500 psi (240 bar)
Particle Size5 μm
pH2.5 to 7.5
Pore Size300 Å
Surface Area100 m²/g
Temperature40°C
Stationary PhaseAmide/Ethyl
Product LineProPac
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Column TypeHydrophobic Interaction
PhaseHydrophobic Interaction
MaterialStainless Steel
Unit SizeEach
Showing 8 of 8
High-efficiency Columns for the Separation of Large Biomolecules
  • High protein loading capacity for protein purification applications
  • Wide range of analytical and preparative applications, including the separation of bovine serum proteins, snake venom proteins, and monoclonal antibodies (Mabs)
  • Based on 5µm ultra-high purity spherical silica gel particles with 300Å pores

Hydrophobic interaction chromatography separates biomolecules in a decreasing salt gradient, based on differences in surface hydrophobicity. This method preserves the biological activity of proteins. The HIC separation mechanism is complementary to those of ion-exchange and gel filtration chromatography.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Figures

Frequently asked questions (FAQs)

Citations & References