Samco™ Wide-Mouth Bio-Tite™ 120mL (4 oz.) 53mm Specimen Containers
Samco™ Wide-Mouth Bio-Tite™ 120mL (4 oz.) 53mm Specimen Containers
Samco™ Wide-Mouth Bio-Tite™ 120mL (4 oz.) 53mm Specimen Containers
Samco™ Wide-Mouth Bio-Tite™ 120mL (4 oz.) 53mm Specimen Containers
Samco™ Wide-Mouth Bio-Tite™ 120mL (4 oz.) 53mm Specimen Containers
Thermo Scientific™

Samco™ Wide-Mouth Bio-Tite™ 120mL (4 oz.) 53mm Specimen Containers

Prevent bio-hazardous spills with leak-resistant wide-mouth containers, ideally suited for urine collection from women during pregnancy and for older or overweight patients.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSterilityClosure ColorLabelNo. per Pack
010051
also known as 01 0051
SterileWhiteNon-tabbed75/Pk., Pre-assembled
010001
also known as 01 0001
SterileOrangeTabbed75/Pk., Pre-assembled
010038
also known as 01 0038
SterileNaturalTabbed75/Pk., Pre-assembled
010100
also known as 01 0100
Non-sterileYellowNon-tabbed75/Pk., Pre-assembled
010101
also known as 01 0101
Non-sterileWhiteNon-tabbed75/Pk., Pre-assembled
010180
also known as 01 0180
Non-sterileNaturalNo Label300/Pk., Bagged Separately
010181
also known as 01 0181
Non-sterileWhiteNo Label300/Pk., Bagged Separately
010182
also known as 01 0182
Non-sterileRedNo Label300/Pk., Bagged Separately
010183
also known as 01 0183
Non-sterileBlueNo Label300/Pk., Bagged Separately
010195
also known as 01 0195
Non-sterileOrangeNo Label300/Pk., Bagged Separately
Catalog number 010051
also known as 01 0051
Price (MXN)
-
Sterility:
Sterile
Closure Color:
White
Label:
Non-tabbed
No. per Pack:
75/Pk., Pre-assembled

Prevent bio-hazardous spills with the proprietary, all plastic, leak-resistant seal of Thermo Scientific™ Samco™ Wide-Mouth Bio-Tite™ 120 mL (4 oz.) 53mm Specimen Containers. Wide-mouth containers are ideally suited for urine collection from women during pregnancy and for older or overweight patients. These stackable, shatter-resistant containers are manufactured from virgin polypropylene and feature the Bio-Tite™ system. They are tested at varying pressures, making them ideal for use in various transport systems.

Features:

  • 120mL (4 oz.) 53mm
  • Bio-Tite caps create leak-resistant seal for superior safety and handling
  • Dual-threaded caps create improved sealing
  • Sterilized by gamma irradiation, which eliminates potential contamination
  • Top and bottom positive grip knurls are easy to grip and open using latex gloves
  • Stackable containers are easy to store, saving lab space
  • Virgin polypropylene containers allow higher clarity to see samples, and will not become brittle
  • Sterile indicator tab assures tamper evidence
  • Bilingual English and Spanish label facilitates accurate usage

Ordering Information:
Case of 300

Specifications
Capacity (Metric)120 mL
Closure ColorWhite
Closure Size53 mm
Closure TypeBio-Tite Lid
DescriptionCap and container pre-assembled, includes sodium thiosulfate tablet
Diameter (Metric)53 mm
LabelNon-tabbed
No. per Case300/Cs.
No. per Pack75/Pk., Pre-assembled
Closure IncludedYes
MaterialVirgin Polypropylene
Product TypeSpecimen Container
ShapeCylindrical
SterilitySterile
Unit SizeCase of 300