Aluminum granules, 8-12mm (0.32-0.47in), 99.9% (metals basis)
Aluminum granules, 8-12mm (0.32-0.47in), 99.9% (metals basis)
Aluminum granules, 8-12mm (0.32-0.47in), 99.9% (metals basis)
Aluminum granules, 8-12mm (0.32-0.47in), 99.9% (metals basis)
Thermo Scientific Chemicals

Aluminum granules, 8-12mm (0.32-0.47in), 99.9% (metals basis)

CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
5 kg
Catalog number 010093.A1
also known as 010093-A1
Price (USD)/ Each
82.65
Online Exclusive
91.60
Save 8.95 (10%)
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
82.65
Online Exclusive
91.60
Save 8.95 (10%)
Add to cart
Chemical Identifiers
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
Molecular Weight (g/mol)26.98
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only