Chromium pieces, irregular, 99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chromium pieces, irregular, 99% (metals basis), Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number 010150.36
also known as 010150-36
Price (USD)/ Each
158.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
187.00 
Save 28.35 (15%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
158.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
187.00 
Save 28.35 (15%)
Add to cart
Chromium pieces, irregular, 99% (metals basis), Thermo Scientific Chemicals
Catalog number010150.36
Price (USD)/ Each
158.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
187.00 
Save 28.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7440-47-3
IUPAC Namechromium
Molecular FormulaCr
InChI KeyVYZAMTAEIAYCRO-UHFFFAOYSA-N
SMILES[Cr]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only