Chromium pieces, electrolytic, 6mm & down, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chromium pieces, electrolytic, 6mm & down, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number 010151.22
also known as 010151-22
Price (USD)/ Each
251.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
251.00
Add to cart
Chromium pieces, electrolytic, 6mm & down, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals
Catalog number010151.22
Price (USD)/ Each
251.00
-
Add to cart
Chemical Identifiers
CAS7440-47-3
IUPAC Namechromium
Molecular FormulaCr
InChI KeyVYZAMTAEIAYCRO-UHFFFAOYSA-N
SMILES[Cr]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Metallic grey
FormPieces, ≤6 mm
Total Metal Impurities≤0.003%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only