Samco™ Wide-Mouth Bio-Tite™ 120mL (4 oz.) 53mm Specimen Containers
Samco™ Wide-Mouth Bio-Tite™ 120mL (4 oz.) 53mm Specimen Containers
Samco™ Wide-Mouth Bio-Tite™ 120mL (4 oz.) 53mm Specimen Containers
Samco™ Wide-Mouth Bio-Tite™ 120mL (4 oz.) 53mm Specimen Containers
Thermo Scientific™

Samco™ Wide-Mouth Bio-Tite™ 120mL (4 oz.) 53mm Specimen Containers

Prevent bio-hazardous spills with leak-resistant wide-mouth containers, ideally suited for urine collection from women during pregnancy and for older or overweight patients.
Have Questions?
變更視圖buttonViewtableView
產品號碼SterilityClosure ColorLabelNo. per Pack
010181
亦稱為 01 0181
Non-sterileWhiteNo Label300/Pk., Bagged Separately
010001
亦稱為 01 0001
SterileOrangeTabbed75/Pk., Pre-assembled
010038
亦稱為 01 0038
SterileNaturalTabbed75/Pk., Pre-assembled
010100
亦稱為 01 0100
Non-sterileYellowNon-tabbed75/Pk., Pre-assembled
010101
亦稱為 01 0101
Non-sterileWhiteNon-tabbed75/Pk., Pre-assembled
010180
亦稱為 01 0180
Non-sterileNaturalNo Label300/Pk., Bagged Separately
010182
亦稱為 01 0182
Non-sterileRedNo Label300/Pk., Bagged Separately
010183
亦稱為 01 0183
Non-sterileBlueNo Label300/Pk., Bagged Separately
010195
亦稱為 01 0195
Non-sterileOrangeNo Label300/Pk., Bagged Separately
產品號碼 010181
亦稱為 01 0181
價格 (HKD)
-
Sterility:
Non-sterile
Closure Color:
White
Label:
No Label
No. per Pack:
300/Pk., Bagged Separately

Prevent bio-hazardous spills with the proprietary, all plastic, leak-resistant seal of Thermo Scientific™ Samco™ Wide-Mouth Bio-Tite™ 120 mL (4 oz.) 53mm Specimen Containers. Wide-mouth containers are ideally suited for urine collection from women during pregnancy and for older or overweight patients. These stackable, shatter-resistant containers are manufactured from virgin polypropylene and feature the Bio-Tite™ system. They are tested at varying pressures, making them ideal for use in various transport systems.

Features:

  • 120mL (4 oz.) 53mm
  • Bio-Tite caps create leak-resistant seal for superior safety and handling
  • Dual-threaded caps create improved sealing
  • Sterilized by gamma irradiation, which eliminates potential contamination
  • Top and bottom positive grip knurls are easy to grip and open using latex gloves
  • Stackable containers are easy to store, saving lab space
  • Virgin polypropylene containers allow higher clarity to see samples, and will not become brittle
  • Sterile indicator tab assures tamper evidence
  • Bilingual English and Spanish label facilitates accurate usage

Ordering Information:
Case of 300

規格
Capacity (Metric)120 mL
Closure ColorWhite
Closure Size53 mm
Closure TypeBio-Tite Lid
DescriptionCap and container bagged separately
Diameter (Metric)53 mm
LabelNo Label
No. per Case300/Cs.
No. per Pack300/Pk., Bagged Separately
Closure IncludedYes
MaterialVirgin Polypropylene
Product TypeSpecimen Container
ShapeCylindrical
SterilityNon-sterile
Unit SizeCase of 300