Gallium ingot, 99.99999% (metals basis)
Gallium ingot, 99.99999% (metals basis)
Gallium ingot, 99.99999% (metals basis)
Gallium ingot, 99.99999% (metals basis)
Thermo Scientific Chemicals

Gallium ingot, 99.99999% (metals basis)

CAS: 7440-55-3 | Ga | 69.72 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
50 g
250 g
500 g
Catalog number 010186.36
also known as 010186-36
Price (EUR)
2.498,00
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (EUR)
2.498,00
Each
Add to cart
Chemical Identifiers
CAS7440-55-3
IUPAC Namegallium
Molecular FormulaGa
InChI KeyGYHNNYVSQQEPJS-UHFFFAOYSA-N
SMILES[Ga]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.00001% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only