Germanium pieces, 3-9mm (0.1-0.4in), 99.999% (metals basis)
Germanium pieces, 3-9mm (0.1-0.4in), 99.999% (metals basis)
Germanium pieces, 3-9mm (0.1-0.4in), 99.999% (metals basis)
Germanium pieces, 3-9mm (0.1-0.4in), 99.999% (metals basis)
Thermo Scientific Chemicals

Germanium pieces, 3-9mm (0.1-0.4in), 99.999% (metals basis)

CAS: 7440-56-4 | Ge | 72.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number 010191.18
also known as 010191-18
Price (USD)/ Each
698.65
Online Exclusive
776.00
Save 77.35 (10%)
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
698.65
Online Exclusive
776.00
Save 77.35 (10%)
Add to cart
Chemical Identifiers
IUPAC Namegermanium
Molecular FormulaGe
InChI KeyGNPVGFCGXDBREM-UHFFFAOYSA-N
SMILES[Ge]
Molecular Weight (g/mol)72.63
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only