Holmium foil, 1.0mm (0.04 in.) thick, 99.9% (REO), Thermo Scientific Chemicals
Holmium foil, 1.0mm (0.04 in.) thick, 99.9% (REO), Thermo Scientific Chemicals
Holmium foil, 1.0mm (0.04 in.) thick, 99.9% (REO), Thermo Scientific Chemicals
Holmium foil, 1.0mm (0.04 in.) thick, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Holmium foil, 1.0mm (0.04 in.) thick, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
010211.FF
also known as 010211-FF
25 x 25 mm
Catalog number 010211.FF
also known as 010211-FF
Price (USD)/ Each
217.65
Special offer
Online exclusive
256.00
Save 38.35 (15%)
Add to cart
Quantity:
25 x 25 mm
Request bulk or custom format
Price (USD)/ Each
217.65
Special offer
Online exclusive
256.00
Save 38.35 (15%)
Add to cart
Chemical Identifiers
IUPAC Nameholmium
Molecular FormulaHo
InChI KeyKJZYNXUDTRRSPN-UHFFFAOYSA-N
SMILES[Ho]
Molecular Weight (g/mol)164.93
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under argon
RUO – Research Use Only