Lutetium foil, 0.5mm (0.02in) thick, 99.9% (REO)
Lutetium foil, 0.5mm (0.02in) thick, 99.9% (REO)
Lutetium foil, 0.5mm (0.02in) thick, 99.9% (REO)
Lutetium foil, 0.5mm (0.02in) thick, 99.9% (REO)
Thermo Scientific Chemicals

Lutetium foil, 0.5mm (0.02in) thick, 99.9% (REO)

CAS: 7439-94-3 | Lu | 174.97 g/mol
Have Questions?
Catalog NumberQuantityPackaging
010229.FF25 x 25 mmMylar under Argon
Catalog number 010229.FF
Price (USD)
852.00
Each
Add to cart
Quantity:
25 x 25 mm
Packaging:
Mylar under Argon
Request bulk or custom format
Price (USD)
852.00
Each
Add to cart
Chemical Identifiers
CAS7439-94-3
IUPAC Namelutetium
Molecular FormulaLu
InChI KeyOHSVLFRHMCKCQY-UHFFFAOYSA-N
SMILES[Lu]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Lutetium is used as an additive for special steel, non-ferric alloy, hydrogen storage alloy, also reducer for making other rare Earth metal. Pipe materials of reactor system. Used in Physical Vapor Deposition (PVD) processes, including thermal and electron-beam (e-beam) evaporation, for the preparation of thin films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
RUO – Research Use Only