Rhenium wire, 0.5mm (0.02in) dia, 99.97% (metals basis)
Rhenium wire, 0.5mm (0.02in) dia, 99.97% (metals basis)
Rhenium wire, 0.5mm (0.02in) dia, 99.97% (metals basis)
Thermo Scientific Chemicals

Rhenium wire, 0.5mm (0.02in) dia, 99.97% (metals basis)

Form: Approx.4.13g/m | CAS: 7440-15-5 | Re | 186.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 cm
25 cm
100 cm
Catalog number 010312.BQ
also known as 010312-BQ
Price (USD)
66.65
Online Exclusive
74.30
Save 7.65 (10%)
Each
Quantity:
5 cm
Request bulk or custom format
Price (USD)
66.65
Online Exclusive
74.30
Save 7.65 (10%)
Each
Chemical Identifiers
CAS7440-15-5
IUPAC Namerhenium
Molecular FormulaRe
InChI KeyWUAPFZMCVAUBPE-UHFFFAOYSA-N
SMILES[Re]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.03% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only