Rubidium, 99.75% (metals basis)
Rubidium, 99.75% (metals basis)
Rubidium, 99.75% (metals basis)
Rubidium, 99.75% (metals basis)
Thermo Scientific Chemicals

Rubidium, 99.75% (metals basis)

CAS: 7440-17-7 | Rb | 85.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 g
5 g
25 g
Catalog number 010315.22
also known as 010315-22
Price (USD)/ Each
2,102.65
Online Exclusive
2,336.00
Save 233.35 (10%)
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
2,102.65
Online Exclusive
2,336.00
Save 233.35 (10%)
Add to cart
Chemical Identifiers
IUPAC Namerubidium
Molecular FormulaRb
InChI KeyIGLNJRXAVVLDKE-UHFFFAOYSA-N
SMILES[Rb]
Molecular Weight (g/mol)85.47
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver-white
Total Metal Impurities≤ 0.25%
FormSolid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only