Titanium powder, -100 mesh, 99.4%
Titanium powder, -100 mesh, 99.4%
Titanium powder, -100 mesh, 99.4%
Titanium powder, -100 mesh, 99.4%
Thermo Scientific Chemicals

Titanium powder, -100 mesh, 99.4%

CAS: 7440-32-6 | Ti | 47.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 010383.36
also known as 010383-36
Price (USD)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS7440-32-6
IUPAC Nametitanium
Molecular FormulaTi
InChI KeyRTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES[Ti]
View more
SpecificationsSpecification SheetSpecification Sheet
Comment-100 mesh
Total Metal Impurities0.6% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only