Tungsten wire, 0.25mm (0.01in) dia, 99.95% (metals basis)
Tungsten wire, 0.25mm (0.01in) dia, 99.95% (metals basis)
Tungsten wire, 0.25mm (0.01in) dia, 99.95% (metals basis)
Tungsten wire, 0.25mm (0.01in) dia, 99.95% (metals basis)
Thermo Scientific Chemicals

Tungsten wire, 0.25mm (0.01in) dia, 99.95% (metals basis)

CAS: 7440-33-7 | W | 183.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 m
10 m
50 m
Catalog number 010408.H7
also known as 010408-H7
Price (USD)/ Each
661.65
Special offer
Online exclusive
778.00
Save 116.35 (15%)
Add to cart
Quantity:
250 m
Request bulk or custom format
Price (USD)/ Each
661.65
Special offer
Online exclusive
778.00
Save 116.35 (15%)
Add to cart
Chemical Identifiers
IUPAC Nametungsten
Molecular FormulaW
InChI KeyWFKWXMTUELFFGS-UHFFFAOYSA-N
SMILES[W]
Molecular Weight (g/mol)183.84
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.05% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only