Tungsten foil, 0.05mm (0.002in) thick, 99.95% (metals basis)
Tungsten foil, 0.05mm (0.002in) thick, 99.95% (metals basis)
Tungsten foil, 0.05mm (0.002in) thick, 99.95% (metals basis)
Thermo Scientific Chemicals

Tungsten foil, 0.05mm (0.002in) thick, 99.95% (metals basis)

CAS: 7440-33-7 | W | 183.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
100 x 100 mm
100 x 500 mm
Catalog number 010417.FI
also known as 010417-FI
Price (EUR)
-
Quantity:
50 x 50 mm
Request bulk or custom format
Chemical Identifiers
CAS7440-33-7
IUPAC Nametungsten
Molecular FormulaW
InChI KeyWFKWXMTUELFFGS-UHFFFAOYSA-N
SMILES[W]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey
FormFoil, 0.05mm (0.002in) thick.
Assay≥99.95% (metals basis)
AssayPb: ≤ 0.002 %
AssaySi: ≤ 0.002 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Frequently asked questions (FAQs)

Is there a coating on the Tungsten foil, 0.05mm (0.002in) thick, 99.95% (metals basis)?

The Tungsten foil, 0.05mm (0.002in) thick, 99.95% (metals basis) may have an oil or oxide coating. To remove the coating, we recommend soaking the foil in very hot water with strong detergent for 10-15 minutes. Wipe the foil with a sponge. Then rinse it with water and dry. This process will remove the coating.