Thermo Scientific Chemicals

Dichlorotris(triphenylphosphine)ruthenium(II), Premion™, 99.95% (metals basis), Ru 10.2% min, Thermo Scientific Chemicals

Catalog number: 010504.06
5 g, Each
Thermo Scientific Chemicals

Dichlorotris(triphenylphosphine)ruthenium(II), Premion™, 99.95% (metals basis), Ru 10.2% min, Thermo Scientific Chemicals

Catalog number: 010504.06
5 g, Each
Quantity
Catalog number: 010504.06
also known as 010504-06
Price (USD)
Price: 260.00
Online price: 220.65
Your price:
Quantity
-

Chemical Identifiers

CAS
15529-49-4
IUPAC Name
λ¹-ruthenium(1+) tris(triphenylphosphane) dichloride
Molecular Formula
C54H45Cl2P3Ru
InChI Key
KSHWMFNJCNGTJO-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Ru+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Total Metal Impurities
0.05% max.

Description

Dichlorotris(triphenylphosphine)ruthenium(II) serves as a precatalyst for the hydrogenation of alkenes, nitro compounds, ketones, carboxylic acids, synthesis of furans from allenyl sulfides and C-H Activation Catalyst in a Ruthenium/Lewis Acid System

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text