Chloropentaammineruthenium(III) chloride, Ru 33.5% min
Chloropentaammineruthenium(III) chloride, Ru 33.5% min
Chloropentaammineruthenium(III) chloride, Ru 33.5% min
Thermo Scientific Chemicals

Chloropentaammineruthenium(III) chloride, Ru 33.5% min

CAS: 18532-87-1 | Cl3H15N5Ru | 292.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 010520.03
also known as 010520-03
Price (USD)
247.65
Special offer
Online exclusive
Ends: 30-Jun-2026
291.00
Save 43.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
247.65
Special offer
Online exclusive
Ends: 30-Jun-2026
291.00
Save 43.35 (15%)
Each
Chemical Identifiers
CAS18532-87-1
IUPAC Nameruthenium(3+) pentaamine trichloride
Molecular FormulaCl3H15N5Ru
InChI KeyHLAVJMYZYQTXAH-UHFFFAOYSA-K
SMILESN.N.N.N.N.[Cl-].[Cl-].[Cl-].[Ru+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Ru 33.5% min
Chloropentaammineruthenium(III) chloride active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only