Thermo Scientific Chemicals

Rhodium(II) acetate, dimer, Premion™, 99.99% (metals basis), Rh 46.2% min, Thermo Scientific Chemicals

Catalog number: 010560.03
1 g, Each
Thermo Scientific Chemicals

Rhodium(II) acetate, dimer, Premion™, 99.99% (metals basis), Rh 46.2% min, Thermo Scientific Chemicals

Catalog number: 010560.03
1 g, Each
Quantity
Catalog number: 010560.03
also known as 010560-03
Price (USD)

Chemical Identifiers

CAS
15956-28-2
IUPAC Name
tetramethyl-2,4,6,8,9,11,12,14-octaoxa-1,5-dirhodatetracyclo[3.3.3.3¹,⁵.0¹,⁵]tetradecane-1,1,5,5-tetrakis(ylium)-3,7,10,13-tetraide
Molecular Formula
C8H12O8Rh2
InChI Key
VOXKIADTKKPBBP-UHFFFAOYSA-N
SMILES
C[C-]1O[Rh++]234O[C-](C)O[Rh++]2(O1)(O[C-](C)O3)O[C-](C)O4
Assay (unspecified)
Rh 46.2% min
Total Metal Impurities
0.01% max.

Description

Catalyst for cyclopropanation of alkenes, oxidation of alcohols, cyclization reactions involving -diazo carbonyl groups, insertion into C-H and X-H bonds (X-H is NH or SH or OH) and for ylide formation. More reactive and useful in differentiating ribonucleosides and deoxynucleosides. It is also used in functionalizing fullerenes into polymers. Efficient catalyst for hydrogen transfer from 2-propanol to cyclohexanone and other unsaturated compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text