Zirconium foil, 1.0mm (0.039in) thick, annealed, 99.2% (metals basis excluding Hf), Hf 4.5% max, Thermo Scientific Chemicals
Zirconium foil, 1.0mm (0.039in) thick, annealed, 99.2% (metals basis excluding Hf), Hf 4.5% max, Thermo Scientific Chemicals
Zirconium foil, 1.0mm (0.039in) thick, annealed, 99.2% (metals basis excluding Hf), Hf 4.5% max, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zirconium foil, 1.0mm (0.039in) thick, annealed, 99.2% (metals basis excluding Hf), Hf 4.5% max, Thermo Scientific Chemicals

Have Questions?
變更視圖buttonViewtableView
Quantity:
50 x 50 mm
100 x 100 mm
100 x 500 mm
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 010590.HP
亦稱為 010590-HP
價格 (TWD)
-
Quantity:
100 x 500 mm
Request bulk or custom format
化學識別
CAS7440-67-7
IUPAC Namezirconium
Molecular FormulaZr
InChI KeyQCWXUUIWCKQGHC-UHFFFAOYSA-N
SMILES[Zr]
檢視更多
規格Specification Sheet規格表
Trace Metal99.2% (metals basis excluding Hf), Hf 4.5% max
TemperAnnealed
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only