Thermo Scientific Chemicals

Aluminum nitrate hydrate, Puratronic™, 99.999% (metals basis excluding Hg), Thermo Scientific Chemicals

Catalog number: 010626.36
500 g, Each
Thermo Scientific Chemicals

Aluminum nitrate hydrate, Puratronic™, 99.999% (metals basis excluding Hg), Thermo Scientific Chemicals

Catalog number: 010626.36
500 g, Each
Quantity
Catalog number: 010626.36
also known as 010626-36
Price (USD)
Price: 811.00
Online price: 702.65
Your price:
Quantity
-

Chemical Identifiers

CAS
25838-59-9
IUPAC Name
aluminium(3+) trinitrate
Molecular Formula
AlN3O9
InChI Key
JLDSOYXADOWAKB-UHFFFAOYSA-N
SMILES
[Al+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Total Metal Impurities
0.001% max.

Description

Aluminum nitrate hydrate is used to produce alumina for the preparation of insulating papers, and in the extraction of actinide elements. It is also used in the preparation of rare earth aluminates, in the heating elements of cathode ray tubes, as a precursor for sol-gel synthesis of alumina, and on transformer core laminates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text